missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC24C Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94294-0.1ml
This item is not returnable.
View return policy
Description
SEC24C Polyclonal antibody specifically detects SEC24C in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| SEC24C | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| KIAA0079SEC24-related protein C, protein transport protein Sec24C, SEC24 (S. cerevisiae) related gene family, member C, SEC24 family, member C (S. cerevisiae), SEC24 related gene family, member C | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 915-1094 of human SEC24C (NP_004913.2). DVLQPGAEVTTDDRAYVRQLVTSMDVTETNVFFYPRLLPLTKSPVESTTEPPAVRASEERLSNGDIYLLENGLNLFLWVGASVQQGVVQSLFSVSSFSQITSGLSVLPVLDNPLSKKVRGLIDSLRAQRSRYMKLTVVKQEDKMEMLFKHFLVEDKSLSGGASYVDFLCHMHKEIRQLLS | |
| 0.1 mL | |
| Signal Transduction | |
| 9632 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction