Learn More
Invitrogen™ SEC61alpha Recombinant Rabbit Monoclonal Antibody (24GB1230)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557247
 View more versions of this product
Product Code. 30276088
  • 500.00 € / 100µL
Description
SEC61alpha Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The SEC61 complex is a major component of the endoplasmic reticulum (ER) translocation site. This complex is composed of three subunits: SEC61alpha, SEC61beta and SEC61gamma. SEC61alpha is a polytopic integral membrane protein. It is a core component of the ER translocation complex, playing a crucial role in the insertion of secretory and membrane polypeptides into the ER.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SEC61alpha
Recombinant Monoclonal
Unconjugated
SEC61A2
protein transport protein Sec61 subunit alpha; protein transport protein Sec61 subunit alpha isoform 2; Sec61 alpha; Sec61 alpha 2 subunit; Sec61 alpha 2 subunit (S. cerevisiae); Sec61 alpha form 2; sec61 alpha-2; Sec61 translocon alpha 2 subunit; Sec61, alpha subunit 2; Sec61, alpha subunit 2 (S. cerevisiae); SEC61A2
Rabbit
Affinity chromatography
RUO
361273, 55176, 57743
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1230
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9H9S3, Q9JLR1
SEC61A2
A synthesized peptide derived from human SEC61A1/2 (aa 350-450).
100 μL
Primary
Human, Mouse, Rat