missing translation for 'onlineSavingsMsg'
Learn More

SecinH3, MedChemExpress

Product Code. 16483078
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483078 10 mg 10mg
16425817 5 mg 5mg
16409646 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16483078 Supplier Medchem Express Supplier No. HY100559
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress SecinH3 is an antagonist of cytohesins with IC50s of 5.4 μM, 2.4 μM, 5.4 μM, 5.6 μM, 5.6 μM and 65 μM for hCyh1, hCyh2, mCyh3, hCyh3, drosophila steppke and yGea2-S7, respectively.

SecinH3 is an antagonist of cytohesins with IC50s of 5.4 μM, 2.4 μM, 5.4 μM, 5.6 μM, 5.6 μM and 65 μM for hCyh1, hCyh2, mCyh3, hCyh3, drosophila steppke and yGea2-S7, respectively.

Specifications

Chemical Name or Material SecinH3
CAS 853625-60-2
Color White
Purity Grade Notes Research
Molecular Formula C24H20N4O4S
Quantity 10 mg
Solubility Information DMSO : 130 mg/mL (282.30 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(N2N=C(OC)N=C2C3=CC=C(OCO4)C4=C3)C=C1)CSC5=CC=CC=C5
Molecular Weight (g/mol) 460.51
Formula Weight 460.51
Percent Purity 99.54%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.