missing translation for 'onlineSavingsMsg'
Learn More

SecinH3, MedChemExpress

Product Code. p-200097568
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16483078

Brand: Medchem Express HY100559

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SecinH3 is an antagonist of cytohesins with IC50s of 5.4 μM, 2.4 μM, 5.4 μM, 5.6 μM, 5.6 μM and 65 μM for hCyh1, hCyh2, mCyh3, hCyh3, drosophila steppke and yGea2-S7, respectively.

SecinH3 is an antagonist of cytohesins with IC50s of 5.4 μM, 2.4 μM, 5.4 μM, 5.6 μM, 5.6 μM and 65 μM for hCyh1, hCyh2, mCyh3, hCyh3, drosophila steppke and yGea2-S7, respectively.

Specifications

Chemical Name or Material SecinH3
CAS 853625-60-2
Color White
Purity Grade Notes Research
Molecular Formula C24H20N4O4S
Quantity 10 mg
Solubility Information DMSO : 130 mg/mL (282.30 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(N2N=C(OC)N=C2C3=CC=C(OCO4)C4=C3)C=C1)CSC5=CC=CC=C5
Molecular Weight (g/mol) 460.51
Formula Weight 460.51
Percent Purity 99.54%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.