Learn More
Erlab Secure Weighing Station Captair 391 Smart
Product Code. 30565337
Click to view available options
Filter Type:
Bonded Carbon and/or HEPA
Unit Size:
Each
Product Code. 30565337
Brand: Erlab AR015
This item requires a price update prior to being ordered.
Request a Price for this item here.
Combines safety, precision, and smart technology for all your sensitive weighing operations. With an anti-vibration slab, recommended for all high-precision weighing, the weighing station ensures reliable, reproducible and stabilized measurements, thereby improving your productivity.
Designed to deliver precision, safety, and user comfort, the 391 Smart secure weighing station meets the highest demands of pharmaceutical, chemical, and industrial applications.
Enhanced Safety
The 391 Smart is fully compliant with AFNOR NF X 15-211:2009 and EN1822-1:2019 standards, offering advanced protection through its HEPA filtration system, maintaining a consistent airflow of up to 300 m³/h. An optional activated carbon filter version (220 m³/h) is also available, ideal for working with volatile substances.
Smart, Connected Technology
With built-in network connectivity, a user-friendly light indicator, and an integrated alarm sensor, the 391 Smart enhances operator safety while simplifying day-to-day operations. Combining an automated balances with the weighing station enables safe handling of powders with varying particle sizes (from granules to nanoparticles) and of liquids such as solvents, acids, and bases.
Premium Work Surface
Equipped with a Trespa™ TopLab™ PLUS surface and a marble insert for precision weighing, this station offers durability, chemical resistance, and easy cleaning, deal for high-accuracy tasks.
Compact and Ergonomic
Dimensions (W x H x D) : 1140 x 1721 x 906 mm (with armrests), 825 mm (without). The oblong sash opening ensures easy access while maintaining maximum containment.
Specifications
Product Type Equipment Containment Enclosure
Filter Type Bonded Carbon and/or HEPA
Material Corrosion Resistant Electro-galvanized Steel, Antiacid Polymer Coating
Voltage 100/240 V