missing translation for 'onlineSavingsMsg'
Learn More

Sedaxane, TRC

Product Code. 17993215
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17993215 250 mg 250mg
17934165 25 mg 25mg
17944835 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17993215 Supplier Toronto Research Chemicals Supplier No. TRCS242000250MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874967-67-6
Molecular Formula C18 H19 F2 N3 O
Molecular Weight (g/mol) 331.36
Synonym N-(2-[1,1'-Bicyclopropyl]-2-ylphenyl)-3-(difluoromethyl)-1-methyl-1H-pyrazole-4-carboxamide, N-(2-Bicyclopropyl-2-ylphenyl)-3-difluoromethyl-1-methyl-1H-pyrazol-4-carboxylic acid amide, N-(2-Bicyclopropyl-2-ylphenyl)-3-difluoromethyl-1-methyl-1H-pyrazole-4-carboxamide, N-[2-(2-(Cyclopropyl)cyclopropyl)phenyl]-3-difluoromethyl-1-methylpyrazole-4-carboxamide, N-[2-(2-Cyclopropylcyclopropyl)phenyl]-3-difluoromethyl-1-methylpyrazole-4-carboxylic acid amide, N-[2-[1,1'-Bi(cyclopropyl)-2-yl]phenyl]-1-methyl-3-(difluoromethyl)-1H-pyrazole-4-carboxamide, Sedaxane, Vibrance
IUPAC Name N-[2-(2-cyclopropylcyclopropyl)phenyl]-3-(difluoromethyl)-1-methylpyrazole-4-carboxamide
SMILES Cn1cc(C(=O)Nc2ccccc2C3CC3C4CC4)c(n1)C(F)F

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C18H19F2N3O/c1-23-9-14(16(22-23)17(19)20)18(24)21-15-5-3-2-4-11(15)13-8-12(13)10-6-7-10/h2-5,9-10,12-13,17H,6-8H2,1H3,(H,21,24)
Formula Weight 331.1496
Percent Purity >95%
Chemical Name or Material Sedaxane
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.