missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEL1L3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-81484
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
SEL1L3 Polyclonal specifically detects SEL1L3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Spécification
| SEL1L3 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| DKFZp781J1697, FLJ21629, KIAA0746FLJ41299, protein sel-1 homolog 3, sel-1 suppressor of lin-12-like 3 (C. elegans), Sel-1L3, Suppressor of lin-12-like protein 3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 23231 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| SEL1L3 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SMNLGYKHYQGIDNYPLDWELSYAYYSNIATKTPLDQHTLQGDQAYVETIRLKDDEILKVQTKEDGDVFMWLKHEATRGNAAAQQRLAQMLFWGQQGVAKNPEAA | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu