missing translation for 'onlineSavingsMsg'
Learn More

Selenium +6 Assurance™ Grade Single Speciation Standard, 125 mL in H2O, SPEX CertiPrep™

Product Code. 11405888 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
125mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11405888 1 Ea. 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11405888 Supplier SPEX Certiprep Supplier No. SPECSE6
  • ONLINE EXCLUSIVE PRICE
    88.00 €
    73.91 € / 125mL
    Save 14.09 €
    16% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Certificate of Analysis

Selenium +6 Assurance™ Grade Single Speciation Standard for sample verification and method validation, 125 mL in H2O, supplied with a comprehensive Certificate of Analysis

  • Selenium +3 Speciation Standard
  • 125 mL volume

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Specifications

Description Speciation +6 Speciation Standard
Packaging EA
CAS 7782-49-2, 7732-18-5
CAS Min % 0.1%, 0.00%
CAS Max % 99.9%, 0.00%
Analyte or Component Names Selenium +6
Molecular Formula Se
Concentration 1000 μg/mL
MDL Number MFCD00134090 MFCD00011224
Technique ICP-MS
UN Number 12352402
Certifications/Compliance ISO/IEC 17025, ISO 17034, ISO 9001
InChI Key BUGBHKTXTAQXES-UHFFFAOYSA-N
Type Certified Reference Material
SMILES [Se]
Element Selenium +6
IUPAC Name selenium
Molecular Weight (g/mol) 78.97
Quantity 1 Ea.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.