missing translation for 'onlineSavingsMsg'
Learn More

Selenium Isotopic Standard: Se @ 10 μg/mL in 2% HNO3 (Enriched in Se-82), VHG™

Product Code. 17994166
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17994166 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17994166 Supplier VHG Labs Supplier No. VHGLIS82SE50
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Selenium Isotopic Standard: Se @ 10 μg/mL in 2% HNO3 (Enriched in Se-82)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Selenium 82
CAS 14687-58-2
Methods ICP-MS
Analyte or Component Names Selenium 82
Shipping Condition Room Temperature
Molecular Formula 82Se
Synonym 82Se
SMILES [82Se]
IUPAC Name Selenium 82
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 81.92
Quantity 50 mL
Formula Weight 81.9167 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.