missing translation for 'onlineSavingsMsg'
Learn More

Seletalisib, MedChemExpress

Product Code. 16413207
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16413207 50 mg 50mg
16497166 100 mg 100mg
16491639 1 mg 1mg
16489707 5 mg 5mg
16487178 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16413207 Supplier Medchem Express Supplier No. HY16754

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Seletalisib (UCB5857) is potent and selective PI3Kδ inhibitor with an IC50 of 12 nM.

Seletalisib (UCB5857) is potent and selective PI3Kδ inhibitor with an IC50 of 12 nM.

Specifications

Chemical Name or Material Seletalisib
Physical Form Solid
CAS 1362850-20-1
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C23H14ClF3N6O
For Use With (Application) COVID-19-immunoregulation
Synonym UCB5857
Solubility Information DMSO : ≥ 83.3 mg/mL (172.52 mM)
SMILES FC(F)(F)[C@H](NC1=C2C(C=CC=N2)=NC=N1)C3=CC4=CC=CC(Cl)=C4N=C3C5=CC=C[N+]([O-])=C5
Molecular Weight (g/mol) 482.85
Formula Weight 482.85
Percent Purity 98.88%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.