missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEMA3F Monoclonal Antibody (741307)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561280
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
The semaphorins are a family of proteins that are involved in signaling. All the family members have a secretion signal, a 500-amino acid sema domain, and 16 conserved cysteine residues (Kolodkin et al., 1993 [PubMed 8269517]). Sequence comparisons have grouped the secreted semaphorins into 3 general classes, all of which also have an immunoglobulin domain. The semaphorin III family, consisting of human semaphorin III (SEMA3A; MIM 603961), chicken collapsin, and mouse semaphorins A, D, and E, all have a basic domain at the C terminus. Chicken collapsin contributes to path finding by axons during development by inhibiting extension of growth cones (Luo et al., 1993 [PubMed 8402908]) through an interaction with a collapsin response mediator protein of relative molecular mass 62K (CRMP62) (Goshima et al., 1995 [PubMed 7637782]), a putative homolog of an axonal guidance associated UNC33 gene product (MIM 601168). SEMA3F is a secreted member of the semaphorin III family.
Specifications
| SEMA3F | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O88632 | |
| SEMA3F | |
| Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 3F, Ala19 Pro775 (Arg583Ala and Arg586Ala). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Frozen) | |
| 741307 | |
| Unconjugated | |
| SEMA3F | |
| LOW QUALITY PROTEIN: semaphorin-3F; sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3 F; sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3F; sema domain, immunoglobulin domain (Ig), short basic domain, secreted, 3F; Sema III/F; Sema IV; Sema3f; Sema4; SemaIV; SEMA-IV; SEMAK; semaphorin 3F; semaphorin III/F; semaphorin IV; Semaphorin-3F | |
| Rat | |
| Protein A/G | |
| RUO | |
| 20350 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction