missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEMA6B Monoclonal Antibody (256537)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561129
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
The SEMA6B gene encodes semaphorin 6B, which belongs to the semaphorin family of proteins known for their roles in guiding axonal development and modulating cellular signaling pathways. Semaphorin 6B primarily functions in the nervous system where it influences axonal guidance, neuronal development, and synapse formation through interactions with neuropilin and plexin receptor complexes. These interactions are crucial during neural development, ensuring proper neuronal wiring and the establishment of functional neural circuits. In addition to its key role in neural connectivity, SEMA6B is increasingly recognized for its involvement in immune responses, where it can modulate the activity and migration of immune cells. In cancer biology, altered expression of SEMA6B has been implicated in tumor progression, affecting tumor cell adhesion and migration, thereby influencing metastasis. Research continues to explore the specific mechanisms of SEMA6B's action and how it can be targeted for therapeutic interventions in neural and cancer-related diseases.
Specifications
| SEMA6B | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O54951 | |
| SEMA6B | |
| Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 6B, Leu27-Ser605 (predicted). | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 256537 | |
| Unconjugated | |
| SEMA6B | |
| Sema; sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6B; sema N; Sema VIB; sema Z; SEMA6B; SEMAN; semaphorin 6B; semaphorin VIB; semaphorin Z; semaphorin-6B; semaphorin-6Ba; semaphorin-N; Semaphorin-Z; SEMA-VIB; semaZ; SEM-SEMA-Y; UNQ1907/PRO4353; VIb | |
| Rat | |
| Protein A/G | |
| RUO | |
| 20359 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction