missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semaphorin 7A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00 € - 624.00 €
Specifications
| Antigen | Semaphorin 7A |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18493311
|
Novus Biologicals
NBP1-86555-25ul |
25 μL |
216.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18291297
|
Novus Biologicals
NBP1-86555 |
0.1 mL |
624.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Semaphorin 7A Polyclonal specifically detects Semaphorin 7A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Semaphorin 7A | |
| Polyclonal | |
| Rabbit | |
| Human | |
| CD108 antigen, CD108MGC126696, CDw108, H-SEMA-K1, H-Sema-L, JMH, JMH blood group antigen, John-Milton-Hargen human blood group Ag, MGC126692, sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin)7A, sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin)7A (JMH blood group), sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, 7A, Sema K1, Sema L, SEMAK1, SEMALJohn Milton Hagen blood group H-Sema K1, semaphorin 7A, GPI membrane anchor (John Milton Hagen blood group), semaphorin K1, semaphorin L, semaphorin-7A, semaphorin-K1, Semaphorin-L | |
| SEMA7A | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 8482 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:QDRVDFGQTEPHTVLFHEPGSSSVWVGGRGKVYLFDFPEGKNASVRTVNIGSTKGSCLDKRDCENYITLLERRSEGLLACGTNARHPSCWNLVNGTVVPLGEMRGYAPFSPDENSLVLFEGD | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title