missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semi Metals, plasma standard solution, Specpure™, As, Bi, Ga, Ge, In, Pb, Sb, Se, Sn, Te, Tl at 100μg/mL
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Semi Metals, plasma standard solution |
| Name Note | As, Bi, Ga, Ge, In, Pb, Sb, Se, Sn, Te, Tl at 100μg/mL |
| Physical Form | Liquid |
| MDL Number | MFCD00145276 |
| UN Number | UN3264 |
| Quantity | 100 mL |
| Grade | Specpure |
Safety and Handling
Product Identifier
- Semi Metals, plasma standard solution, Specpure®, As, Bi, Ga, Ge, In, Pb, Sb, Se, Sn,Te, Tl @ 100myg/ml
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Hydrochloric acid, Hydrofluoric acid
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction