missing translation for 'onlineSavingsMsg'
Learn More

Sennoside Calcium Salt (Mixture of A and B), TRC

Product Code. 30412335
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412335 50 mg 50mg
30426234 100 mg 100mg
30419422 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30412335 Supplier Toronto Research Chemicals Supplier No. TRCS25880050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sennoside Calcium Salt (Mixture of A and B)
Analyte or Component Names Sennoside Calcium Salt (Mixture of A and B)
Shipping Condition Room Temperature
Molecular Formula C42 H36 O20 . Ca
Synonym (9R,9'RS)-5,5'-Bis(β-D-glucopyranosyloxy)-9,9',10,10'-tetrahydro-4,4'-dihydroxy-10,10'-dioxo-[9,9'-bianthracene]-2,2'-dicarboxylic Acid Calcium Salt
InChI Formula InChI=1S/C42H38O20.Ca/c43-11-23-31(47)35(51)37(53)41(61-23)59-21-5-1-3-15-25(17-7-13(39(55)56)9-19(45)27(17)33(49)29(15)21)26-16-4-2-6-22(60-42-38(54)36(52)32(48)24(12-44)62-42)30(16)34(50)28-18(26)8-14(40(57)58)10-20(28)46;/h1-10,23-26,31-32,35-38,41-48,51-54H,11-12H2,(H,55,56)(H,57,58);/q;+2/p-2/t23-,24-,25?,26?,31-,32-,35+,36+,37-,38-,41-,42-;/m1./s1
Solution Type Neat
SMILES [Ca+2].OC[C@H]1O[C@@H](Oc2cccc3C(C4c5cccc(O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)c5C(=O)c7c(O)cc(cc47)C(=O)[O-])c8cc(cc(O)c8C(=O)c23)C(=O)[O-])[C@H](O)[C@@H](O)[C@@H]1O
IUPAC Name calcium;9-[2-carboxylato-4-hydroxy-10-oxo-5-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-9H-anthracen-9-yl]-4-hydroxy-10-oxo-5-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-9H-anthracene-2-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 900.801
Quantity 50 mg
Formula Weight 900.143
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.