missing translation for 'onlineSavingsMsg'
Learn More

SENP2 Antibody (SR2130), Novus Biologicals™

Rabbit Monoclonal Antibody

338.00 € - 449.00 €

Specifications

Antigen SENP2
Clone SR2130
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18671824
View Documents
Novus Biologicals
NBP3-22280-100ul
100 μg
449.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18631124
View Documents
Novus Biologicals
NBP3-22280-50ul
50 μg
338.00 €
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SENP2 Monoclonal antibody specifically detects SENP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications

Specifications

SENP2
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Monoclonal
Purified
RUO
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
59343
IgG
Affinity purified
SR2130
Western Blot, Immunohistochemistry, Immunofluorescence
Unconjugated
Rabbit
Human
AXAM2Axam2, DKFZp762A2316, EC 3.4.22, EC 3.4.22.68, KIAA1331sentrin-specific protease 2, Sentrin/SUMO-specific protease SENP2, Smt3ip2, SMT3IP2EC 3.4.22.-, SMT3-specific isopeptidase 2, SUMO1/sentrin/SMT3 specific peptidase 2, SUMO1/sentrin/SMT3 specific protease 2
A synthesized peptide derived from human SENP2 (Uniprot #: Q9HC62)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.