Learn More
Description
Range of weak and strong anionic (A) and cationic (C) exchangers. Fractionation range: A-25 and C-25, up to 30 000; A-50 and C-50: 30 000–100 000.
Very high capacities even at elevated ionic strengths
Very low non-specific adsorption
Wide range of applications at laboratory and process scales
Suitable for column or batch techniques
Simple and economical to use
DEAE Sephadex
- Weak and strong anion exchanger
- pH range 2-9
QAE Sephadex
- Strong anion exchanger
- pH range: 2-12
- Weak cation exchanger
- pH range: 6-10
- Strong cation exchanger
- pH range: 4-13
Spécification
Spécification
| Quantity | 100 g |
| Particle Size | 40 to 120 μm in dry form |
| For Use With (Equipment) | Column or Batch Techniques |
| Product Type | Sephadex Ion Exchange Media |
| Format | Solid Spherical Bead |
| For Use With (Application) | Separating and purifying Proteins, Polypeptides, Nucleic Acids and other charged biomolecules by using column or batch techniques |
| Height (English) | 2 to 5.9 in. |
| Height (Metric) | 5 to 15cm |
| Ligand Type | Carboxymethyl |
| Material | CM-Sephadex C-25 |
| Afficher plus |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.