missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sepharose Agarose Gel Filtration Media

Choose from a wide range of bead-formed agarose-based gel filtration media. Cytiva Life Sciences™ Sepharose Agarose Gel Filtration Media shows good separation over a wide molecular weight range.

Brand:  Cytiva 17015001

Code : GE

Additional Details : Weight : 1.43100kg

 View more versions of this product

Product Code. 10004634

  • 1059.00 € / 1 L
Estimated Shipment: 05-06-2024
to see stock.
Add to basket

Description

Description

European Pharmacopoeia Solutions
  • Reagents and Standard solutions in Chapters 2 and 4 of the current Ph. Eur.
  • All are manufactured and tested in compliance with the Ph. Eur.
  • Supplied with a Certificates of Analysis
  • Lot No. and expiry date are stated on all product labels
Specifications

Specifications

1L
45 to 165 μm
Solid Bead
Allow characterization or cleaning up of samples containing components of diverse molecular weight
10cm
3 to 13
4 to 30°C, 20% Ethanol
<26 cm/hr.
Sepharose Agarose Gel Filtration Media
5cm
3.94 in.
80-150 cm/h, pressure drop cm H2O/bed height=15, bed height 10 cm, column 5 cm i.d.
2% Agarose
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers