missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Septin 2 Recombinant Rabbit Monoclonal Antibody (24GB1525)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557709
This item is not returnable.
View return policy
Description
Septin 2 Recombinant Monoclonal Antibody for Western Blot, Flow
Septin 2 is a filament-forming cytoskeletal GTPase. Septin 2 is required for normal organization of the actin cytoskeleton and plays a role in the biogenesis of polarized columnar-shaped epithelium by maintaining polyglutamylated microtubules, thus facilitating efficient vesicle transport, and by impeding MAP4 binding to tubulin. It is required for the progression through mitosis, and it forms a scaffold at the midplane of the mitotic splindle required to maintain CENPE localization at kinetochores and consequently chromosome congression. During anaphase, Septin 2 may be required for chromosome segregation and spindle elongation. Septin 2 also plays a role in ciliogenesis and collective cell movements.
Specifications
| Septin 2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| SEPTIN2 | |
| 2-Sep; AW208991; DIFF6; hNedd5; KIAA0158; mKIAA0158; NEDD5; Nedd-5; Neural precursor cell expressed developmentally down-regulated protein 5; neural precursor cell expressed, developmentally down-regulated 5; neural precursor cell expressed, developmentally down-regulated gene 5; Pnutl3; Sept2; septin 2; Septin2; septin-2; Vascular endothelial cell specific protein 11; Vesp11 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 117515, 18000, 4735 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1525 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P42208, Q15019, Q91Y81 | |
| SEPTIN2 | |
| A synthesized peptide derived from human Septin 2 (aa 250-300). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction