missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Septin-5, Mouse anti-Human,Rat,Rabbit, Clone: SP18, Novus Biologicals™
Mouse Monoclonal Antibody
250.00 € - 655.00 €
Specifications
| Antigen | Septin-5 |
|---|---|
| Clone | SP18 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, ELISA, Immunohistochemistry |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18228859
|
Novus Biologicals
NBP2-81058-0.025MG |
0.025 mg |
250.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18394110
|
Novus Biologicals
NBP2-81058-0.2MG |
0.2 mg |
655.00 €
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Septin-5 Monoclonal specifically detects Septin-5 in Human, Rat, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
| Septin-5 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication | |
| PBS | |
| 5413 | |
| Immunoprecipitate from human brain. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| SP18 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat, Rabbit | |
| CDCREL, CDCREL1, CDCREL-1, cell division control related protein 1, Cell division control-related protein 1, HCDCREL-1, peanut-like 1, peanut-like 1 (Drosophila), Peanut-like protein 1, platelet glycoprotein Ib beta chain, PNUTL1H5, septin 5, septin-5 | |
| SEPTIN5 | |
| IgG1 κ | |
| Protein A purified | |
| This antibody recognises Septin-5 (CDCrel-1) and does not cross-react with CDCrel-2. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title