missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERBP1 Monoclonal Antibody (OTI2E11), TrueMAB™, OriGene
Description
SERBP1 may play a role in the regulation of mRNA stability. It binds to the 3′-most 134 nt of the SERPINE1/PAI1 mRNA, a region which confers cyclic nucleotide regulation of message decay.
Specifications
Specifications
| Antigen | SERBP1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI2E11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | SERBP1 |
| Gene Accession No. | Q6AXS5, Q8NC51, Q9CY58 |
| Gene Alias | CGI-55, CHD3IP, HABP4L, PAI-RBP1, PAIRBP1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?