missing translation for 'onlineSavingsMsg'
Learn More

Scientific Industries Serial Cable for Enviro-Genie

Product Code. 11371454 Shop All Ohaus Products
Change view
Click to view available options
For Use With (Equipment):
Enviro-Genie
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11371454 Enviro-Genie Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11371454 Supplier Scientific Industries Supplier No. 83041375
  • ONLINE EXCLUSIVE PRICE
    28.35 €
    24.03 € / Pack of 1
    Save 4.32 €
    15% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Serial Cable may be used to connect the Enviro-Genie to a computer via a serial port for data capture or programming.

  • Used to connect the Enviro-Genie to a computer via a serial port for data capture or programming
  • Standard 9-pin, female-to-female straight through serial cable
TRUSTED_SUSTAINABILITY

Specifications

Product Type Serial Cable
Weight (English) 1 lb.
Weight (Metric) 0.45 kg
Color Gray
For Use With (Application) Data transfer and programming via serial port
For Use With (Equipment) Enviro-Genie
Packaging Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.