Learn More
SERINC5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-91164
This item is not returnable.
View return policy
Description
SERINC5 Polyclonal specifically detects SERINC5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SERINC5 | |
| Polyclonal | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:AHKMKEHIPFFEDMCKGIKAGDTCEKLV | |
| RUO | |
| IgG |
| Immunohistochemistry | |
| Rabbit | |
| 0.1 mL | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |