missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin E1/PAI-1 Antibody (SR1779), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00 € - 449.00 €
Specifications
| Antigen | Serpin E1/PAI-1 |
|---|---|
| Clone | SR1779 |
| Dilution | Western Blot 1:500-1:1000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:30 |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18662674
|
Novus Biologicals
NBP3-21747-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18649933
|
Novus Biologicals
NBP3-21747-50ul |
50 μg |
338.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serpin E1/PAI-1 Monoclonal antibody specifically detects Serpin E1/PAI-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, ImmunoprecipitationSpecifications
| Serpin E1/PAI-1 | |
| Western Blot 1:500-1:1000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:30 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Endothelial plasminogen activator inhibitor, PAI1PAI-1, PAISerpin E1, PLANH1plasminogen activator inhibitor 1, serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 1, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 1 | |
| A synthesized peptide derived from human Serpin E1/PAI-1 (Uniprot #: P05121) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1779 | |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 5054 | |
| IgG | |
| Affinity purified |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto