missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERPINB4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145061
This item is not returnable.
View return policy
Description
Immunogen sequence: DLKDTLRTMG MVNIFNGDAD LSGMT Antigen sequence identity: Mouse 54%; Rat 54%.
SERPINB4 is also called squamous cell carcinoma antigen 2 (SCCA-2) or leupin. It directly inhibits human granzyme proteolytic activity and that overexpression of SERPINB4 in HeLa cells inhibits recombinant granzyme-induced as well as NK cell-mediated cell death.
Specifications
| SERPINB4 | |
| Polyclonal | |
| Unconjugated | |
| SERPINB4 | |
| 1110001H02Rik; 1110013A16Rik; LEUPIN; Leupin SERPINB4; Peptidase inhibitor 11; PI11; PI-11; protease inhibitor (leucine-serpin); RGD1562868; SCCA1; SCCA2; SCCA-2; SCCA2/SCCA1 fusion protein; serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 3A; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3A; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 4; serine peptidase inhibitor, clade B, member 3A; serpin B4; serpin family B member 4; serpin peptidase inhibitor, clade B (ovalbumin), member 4; Serpinb3a; SERPINB4; Sqn5; squamous cell carcinoma antigen 1; squamous cell carcinoma antigen 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6318 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P48594 | |
| SERPINB4 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 294-318. Recombinant protein control fragment (Product #RP-109760). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction