missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SERPINB9 Monoclonal Antibody (7D8)
GREENER_CHOICE

Product Code. 15353931
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15353931 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15353931 Supplier Invitrogen™ Supplier No. MA517648

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA5-17648 targets PI-9/Proteinase Inhibitor 9 in ELISA, FACS, and WB applications and shows reactivity with Human samples. The MA5-17648 immunogen is recombinant PI-9 produced in P. pastoris.

PI9 belongs to the large superfamily of serine proteinase inhibitors (serpins), which bind to and inactivate serine proteinases. These interactions are involved in many cellular processes, including coagulation, fibrinolysis, complement fixation, matrix remodeling, and apoptosis (Sprecher et al., 1995 [PubMed 8530382]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINB9
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 7D8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene SERPINB9
Gene Accession No. P50453
Gene Alias BB283241; CAP3; CAP-3; Cytoplasmic antiproteinase 3; ovalbumin; peptidase inhibitor 9; PI9; PI-9; protease inhibitor 9 (ovalbumin type); serine (or cysteine) peptidase inhibitor, clade B, member 9; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 9; serine (or cysteine) proteinase inhibitor, clade B, member 9; serine protease inhibitor 6; serpin B9; serpin family B member 9; serpin peptidase inhibitor, clade B (ovalbumin), member 9; serpin peptidase inhibitor, clade B, member 9; SERPINB9; Spi6; testicular tissue protein Li 180
Gene Symbols SERPINB9
Host Species Mouse
Immunogen Recombinant PI-9 produced in P. pastoris.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5272
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.