missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serum Amyloid A1/A2 Antibody (SR1117), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21727-100ul
This item is not returnable.
View return policy
Description
Serum Amyloid A1/A2 Monoclonal antibody specifically detects Serum Amyloid A1/A2 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Serum Amyloid A1/A2 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6288 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| SR1117 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| PIG4;SAA;SAA1;SAA2;Serum amyloid A-1 protein;Serum amyloid A-2 protein;TP53I4 | |
| A synthesized peptide derived from human Serum Amyloid A1/A2 (Uniprot #: P0DJI8) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction