Learn More
Miele Professional™ Service Lab Washer - Requalification
Product Code. 30764752 Shop All Miele Products
Change view
Click to view available options
For Use With (Equipment):
Lab Washer
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
30764752 Lab Washer Each
1 options
Product Code. 30764752 Supplier Miele Professional™ Supplier No. 12339840
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Regular inspections to ensure process security with calibration and operational/performance verification
  • Miele Professional's Requalification service typically encompasses two main components that may be offered individually or together: Calibration and Operational/Performance Verification
  • The process of checking and adjusting the machine's measuring systems and componentry to ensure accuracy. This is often an Annual Maintenance and Calibration service
  • A set of documented tests to demonstrate that the machine continues to function as designed and can consistently operate within established limits. This verifies machine performance after a period of use
  • Provides documented proof that the unit achieves the necessary thermal disinfection or cleaning temperature profiles required by regulatory standards.
  • The service includes the creation of a formal, detailed protocol (report) that documents the tests performed, the results, and any adjustments made
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Lab Washer
Product Type Requalification Service