missing translation for 'onlineSavingsMsg'
Learn More

Sesamol, TRC

Product Code. 30427381
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30427381 5 g 5g
30412057 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30427381 Supplier Toronto Research Chemicals Supplier No. TRCS2806005G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 533-31-3
Molecular Formula C7 H6 O3
Molecular Weight (g/mol) 138.12
Synonym Sesamol, 1,3-Benzodioxol-5-ol, Phenol, 3,4-(methylenedioxy)- (7CI,8CI), Sesamol (6CI), 3,4-(Methylenedioxy)phenol, 4-Hydroxy-1,2-methylenedioxybenzene, 5-Hydroxy-1,3-benzodioxole, NSC 59256
IUPAC Name 1,3-benzodioxol-5-ol
SMILES Oc1ccc2OCOc2c1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H6O3/c8-5-1-2-6-7(3-5)10-4-9-6/h1-3,8H,4H2
Formula Weight 138.0317
Chemical Name or Material Sesamol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.