missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SESN3 Native Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
0.15 mL
Unit Size:
150µL
Specifications
Specifications
| For Use With (Application) | Western Blot |
| Formulation | SDS PAGE sample buffer |
| Gene ID (Entrez) | 143686 |
| Quantity | 0.15 mL |
| Storage Requirements | Store at -20°C. Avoid freeze/thaw cycles. |
| Gene Alias | SEST3MGC29667, sestrin 3, sestrin-3 |
| Gene Symbol | SESN3 |
| Inhibitor Properties | Western blotting positive control for SESN3 |
| Research Category | Cell Cycle and Replication |
| Purity or Quality Grade | >85%, by HPLC |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction