missing translation for 'onlineSavingsMsg'
Learn More

Set of 3 QC Check Samples in PE Powder: Br, Cd, Cr, Hg, Pb

Product Code. 30316647
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316647 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316647 Supplier VHG Labs Supplier No. VHGROHSPESET1P
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromine∣Cadmium∣Chromium∣Lead∣Mercury
CAS 7726-95-6, 7440-43-9, 7440-47-3, 7439-92-1, 7439-97-6
Analyte or Component Names Bromine∣Cadmium∣Chromium∣Lead∣Mercury
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solution Type Matrix Material
Element Multi-element
Solvent or Matrix Polyethylene
Industry Type Petrochemical Testing
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.