missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETD3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
280.00 € - 572.00 €
Specifications
| Antigen | SETD3 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18444701
|
Novus Biologicals
NBP1-88416-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18243447
|
Novus Biologicals
NBP1-88416 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SETD3 Polyclonal specifically detects SETD3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| SETD3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 84193 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ERASPNSFWQPYIQTLPSEYDTPLYFEEDEVRYLQSTQAIHDVFSQYKNTARQYAYFYKVIQTHPHANKLPLKDSFTYEDYRWAVSSVMTRQNQIPTEDGSR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| C14orf154, chromosome 14 open reading frame 154, DKFZp761E1415, EC 2.1.1.43, FLJ23027, MGC87236, SET domain containing 3, SET domain-containing protein 3 | |
| SETD3 | |
| IgG | |
| Affinity Purified | |
| Specificity of human SETD3 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title