missing translation for 'onlineSavingsMsg'
Learn More

Sethoxydim 50 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30314983
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30314983 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314983 Supplier Dr. Ehrenstorfer Supplier No. DREA16940000AL50
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sethoxydim
CAS 74051-80-2
Analyte or Component Names Sethoxydim
Shipping Condition +20°C
Molecular Formula C17 H29 N O3 S
Concentration 50 μg/mL
Certifications/Compliance ISO 17034
Synonym 2-[1-(Ethoxyimino)butyl]-5-[2-(ethylthio)propyl]-3-hydroxy-2-cyclohexen-1-one, ARD 34/02, Alloxol S, BAS 9052, BAS 90520H, BASF 9052, Cyethoxydim, Expand, Expand Plus, Fervinal, Fervinal plus, Grasidim, NP 55, Nabu, Nabu S, Poast, Poast Plus, Segment, Sertin, Sethoxydim
InChI Formula InChI=1S/C17H29NO3S/c1-5-8-14(18-21-6-2)17-15(19)10-13(11-16(17)20)9-12(4)22-7-3/h12-13,19H,5-11H2,1-4H3
SMILES CCCC(=NOCC)C1=C(O)CC(CC(C)SCC)CC1=O
IUPAC Name 2-(N-ethoxy-C-propylcarbonimidoyl)-5-(2-ethylsulfanylpropyl)-3-hydroxycyclohex-2-en-1-one
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 327.48
Quantity 1 mL
Formula Weight 327.1868 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.