missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SF2 Rabbit anti-Human, Mouse, Rat, Clone: 8L3T3, Novus Biologicals™
Rabbit Monoclonal Antibody
201.00 € - 510.00 €
Specifications
| Antigen | SF2 |
|---|---|
| Clone | 8L3T3 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18345464
|
Novus Biologicals
NBP3-16329-20UL |
20 μg |
201.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18324955
|
Novus Biologicals
NBP3-16329-100UL |
100 μg |
510.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SF2 Monoclonal antibody specifically detects SF2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| SF2 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Alternative-splicing factor 1, ASFpre-mRNA-splicing factor SF2, P33 subunit, MGC5228, serine/arginine-rich splicing factor 1, SF2FLJ53078, SF2p33, SFRS1splicing factor 2, Splicing factor, arginine/serine-rich 1ASF-1, SR splicing factor 1, SRp30a | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SF2 (Q07955). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 8L3T3 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| DNA replication Transcription Translation and Splicing | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 6426 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title