missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SF3A2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00 € - 624.00 €
Specifications
| Antigen | SF3A2 |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
|
18664379
|
Novus Biologicals
NBP2-48775-25ul |
25 μL |
415.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18616485
|
Novus Biologicals
NBP2-48775 |
0.1 mL |
624.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivning
SF3A2 Polyclonal antibody specifically detects SF3A2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifikationer
| SF3A2 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| pre-mRNA splicing factor SF3A, subunit 2, PRP11, PRPF11, SAP 62, SAP62Prp11, SF3a66splicing factor 3a, subunit 2, 66kD, spliceosome associated protein 62, Spliceosome-associated protein 62, splicing factor 3A subunit 2, splicing factor 3a, subunit 2, 66kDa | |
| This antibody was developed against a recombinant protein corresponding to amino acids: EVKKFVKIGRPGYKVTKQRDSEMGQQSLLFQIDYPEIAEGIMPRHRFMSAYEQRIEPPDRRWQYLLMAAEPYETIAFKVPSREIDKAEGKFWTHWNRETKQFFLQFHFKMEKP | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2), 40% Glycerol | |
| 8175 | |
| IgG | |
| Immunogen affinity purified |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel