Learn More
SF3A3 Antibody (S06-3I7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19809-50ul
 View more versions of this product
Product Code. 18656853
  • 421.00 € / 50µL
Description
SF3A3 Monoclonal antibody specifically detects SF3A3 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SF3A3
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
10946
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot
S06-3I7
Unconjugated
PRP9, PRPF9, SAP 61, SAP61splicing factor 3a, subunit 3, 60kD, SF3a60pre-mRNA splicing factor SF3a (60kD), spliceosome associated protein 61, Spliceosome-associated protein 61, splicing factor 3A subunit 3, splicing factor 3a, subunit 3, 60kDa
A synthetic peptide of human SF3A3 (Uniprot # Q12874)
50 μg
Primary
Human, Mouse, Rat
Purified