Learn More
SF3B3 Rabbit, anti-Human, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL594-14577-20UL
 View more versions of this product
Product Code. 18056875
  • 140.00 € / 20µL
Description
The SF3B3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SF3B3. This antibody recognizes human, mouse, rat antigen. The SF3B3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
Specifications
SF3B3
1000 μg/mL
KIAA0017, RSE1, SAP 130, SAP130, SF3b130, SF3B3, Splicing factor 3B subunit 3, STAF130
20 μL
23450
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite 594
Rabbit
Primary
Human, Mouse, Rat
150 kDa