missing translation for 'onlineSavingsMsg'
Learn More

SF3B3 Rabbit, anti-Human, CoraLite 594, Polyclonal, Proteintech

Product Code. 18060296 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18060296 100 μL 100µL
18056875 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18060296 Supplier Proteintech Supplier No. CL59414577100UL
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The SF3B3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SF3B3. This antibody recognizes human, mouse, rat antigen. The SF3B3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.

Specifications

Antigen SF3B3
Applications Immunofluorescence
Concentration 1000 μg/mL
Conjugate CoraLite 594
Gene Symbols KIAA0017, RSE1, SAP 130, SAP130, SF3b130, SF3B3, Splicing factor 3B subunit 3, STAF130
Host Species Rabbit
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 23450
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.