Learn More
SFRS11 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26751-1-AP-150UL
Description
The SFRS11 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SFRS11. This antibody recognizes human, rat antigen. The SFRS11 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| SFRS11 | |
| 250 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| p54, SFRS11, SRSF11 | |
| 150 μL | |
| 9295 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |