missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFRS15 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-88003
This item is not returnable.
View return policy
Description
SFRS15 Polyclonal specifically detects SFRS15 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SFRS15 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| CTD-binding SR-like protein RA4, FLJ23364, KIAA1172arginine/serine-rich 15, SFRS15, SR-like CTD-associated factor 4, SR-related and CTD-associated factor 4, SR-related CTD-associated factor 4 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 57466 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| SCAF4 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KVKPEELESFCEGGMLDSDTLNPDWKGIPKKPENEVAQNGGAETSHTEPVSPIPKPLPVPVPPIPVPAPITVPPPQVPPHQPGPPVVGALQP | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction