missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFRS8 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94834-0.1ml
This item is not returnable.
View return policy
Description
SFRS8 Polyclonal antibody specifically detects SFRS8 in Human samples. It is validated for Western Blot
Specifications
| SFRS8 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Drosophila homolog), Drosophila), SFRS8, Splicing factor, arginine/serine-rich 8, splicing factor, arginine/serine-rich 8 (suppressor-of-white-apricot, splicing factor, arginine/serine-rich 8 (suppressor-of-white-apricot homolog, splicing factor, suppressor of white-apricot homolog, splicing factor, suppressor of white-apricot homolog (Drosophila), Suppressor of white apricot protein homolog, SWAPMGC167082 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 170-280 of human SFSWAP (NP_004583.2). SKQREKNEAENLEENEEPFVAPLGLSVPSDVELPPTAKMHAIIERTASFVCRQGAQFEIMLKAKQARNSQFDFLRFDHYLNPYYKFIQKAMKEGRYTVLAENKSDEKKKSG | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6433 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction