missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFRS9 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94886-0.02ml
This item is not returnable.
View return policy
Description
SFRS9 Polyclonal antibody specifically detects SFRS9 in Human, Mouse samples. It is validated for Western Blot
Specifications
| SFRS9 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Pre-mRNA-splicing factor SRp30C, serine/arginine-rich splicing factor 9, SFRS9, Splicing factor, arginine/serine-rich 9SR splicing factor 9, SRp30c | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-221 of human SFRS9 (NP_003760.1). MSGWADERGGEGDGRIYVGNLPTDVREKDLEDLFYKYGRIREIELKNRHGLVPFAFVRFEDPRDAEDAIYGRNGYDYGQCRLRVEFPRTYGGRGGWPRGGRNGPPTRRSDFRVLVSGLPPSGSWQDLKDHMREAGDVCYADVQKDGVGMVEYLRKEDMEYALRKLDDTKFRSHEGETSYIRVYPERSTSYGYSRSRSGSRGRDSPYQSRGSPHYFSPFRPY | |
| 0.02 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 8683 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction