missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFTS Virus HB29 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP2-41153-0.025mg
This item is not returnable.
View return policy
Description
SFTS Virus HB29 Polyclonal specifically detects SFTS Virus HB29 in Virus samples. It is validated for Western Blot, ELISA.
Specifications
| SFTS Virus HB29 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| SFTS Virus HB29 Membrane Glycoprotein antibody is specific to the SFTS Virus HB29. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot, ELISA 1 μg/mL | |
| B303_sMgp1, Membrane glycoprotein polyprotein | |
| Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human SFTS Virus HB29 Membrane Glycoprotein. The immunogen is located within amino acids 60 - 110 of SFTS Virus HB29. Amino Acid Squence: QRGGRSQVSYYPAENSYSR | |
| 0.025 mg | |
| Primary | |
| Virus | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction