Learn More
Trajan™ BPX35 GC Capillary Column
Contains 35% Phenyl Polysilphenylene-siloxane. Trajan Scientific and Medical™ BPX35 Capillary GC Column is an MS premium, low bleed phase designed for trace analysis of herbicides and aromatics.
426.00 € - 1180.00 €
Product Code | Brand | Diameter (Metric) Inner | Film Thickness | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Film Thickness | Length (Metric) | Price | Quantity & Availability | |||
15208703
|
Trajan™
054699 |
0.1 mm | 0.1 μm | 10 m |
426.00 €
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||
15238703
|
Trajan™
054737 |
0.53 mm | 1 μm | 30 m |
835.00 €
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||
15228703
|
Trajan™
0547158 |
0.32 mm | 0.5 μm | 30 m |
730.00 €
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||
12945551
|
Trajan™
054702 |
0.25 mm | 0.25 μm | 60 m |
1180.00 €
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||
10102882
|
Trajan™
054701 |
0.25 mm | 0.25 μm | 30 m |
695.00 €
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||
10626211
|
Trajan™
054711 |
0.22 mm | 0.25 μm | 25 m |
785.00 €
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||
10075400
|
Trajan™
054724 |
0.32 mm | 0.25 μm | 30 m |
730.00 €
Each |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||
Description
- Mid-polarity column
- Ideal for confirmational analysis
- High temperature
- Low bleed
- Ideal for drugs of abuse, pharmaceuticals, PAHs, pesticides and herbicides
Specifications
10°C to 330/360°C | |
Mid polar |