missing translation for 'onlineSavingsMsg'
Learn More

SGN-2FF, MedChemExpress

Product Code. 16490467
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16490467

Brand: Medchem Express HY107366

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SGN-2FF is a potent and orally active inhibitor of fucosylation, directly inhibits fucosyltransferase activity. SGN-2FF possesses antitumor activity.

SGN-2FF is a potent and orally active inhibitor of fucosylation, directly inhibits fucosyltransferase activity. SGN-2FF possesses antitumor activity.

Specifications

Chemical Name or Material SGN-2FF
Physical Form Solid
CAS 2089647-47-0
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C6H11FO4
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 50 mg/mL (300.93 mM; Need ultrasonic) ∣H2O : 36.67 mg/mL (220.70 mM; Need ultrasonic)
SMILES F[C@H]1[C@@H]([C@@H]([C@H](C)OC1O)O)O
Molecular Weight (g/mol) 166.15
Formula Weight 166.15
Percent Purity 99.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.