missing translation for 'onlineSavingsMsg'
Learn More

SGX-523, MedChemExpress

Product Code. 16480146
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16480146 2 mg 2mg
16425706 100 mg 100mg
16414928 10 mg 10mg
16407677 5 mg 5mg
16401177 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16480146 Supplier Medchem Express Supplier No. HY12019
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress SGX523 is a exquisitely selective and ATP-competitive MET inhibitor. SGX523 potently inhibits MET with an IC50 of 4 nM and is >1,000-fold selective versus other protein kinases. Antitumor activity.

SGX523 is a exquisitely selective and ATP-competitive MET inhibitor. SGX523 potently inhibits MET with an IC50 of 4 nM and is >1,000-fold selective versus other protein kinases. Antitumor activity.

Specifications

Chemical Name or Material SGX-523
Physical Form Solid
CAS 1022150-57-7
Color Light Yellow
Quantity 2 mg
Purity Grade Notes Research
Molecular Formula C18H13N7S
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 30 mg/mL (83.47 mM; Need ultrasonic)
SMILES CN1N=CC(C2=NN3C(SC4=CC5=C(N=CC=C5)C=C4)=NN=C3C=C2)=C1
Molecular Weight (g/mol) 359.41
Formula Weight 359.41
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.