missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ SH2D1A Antibody (13) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

195.00 € - 361.00 €

Specifications

Antigen SH2D1A
Clone 13
Dilution ELISA 1:1000-1:2000, Sandwich ELISA Detection 1:1000-1:10000
Applications ELISA, Sandwich ELISA Detection
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043454
View Documents
Novus Biologicals™
NBP3-26885-100ul
100 μL
361.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043976
View Documents
Novus Biologicals™
NBP3-26885-20ul
20 μL
195.00 €
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SH2D1A Monoclonal antibody specifically detects SH2D1A in Human samples. It is validated for ELISA,Sandwich ELISA Detection
Specifications

Specifications

SH2D1A
ELISA 1:1000-1:2000, Sandwich ELISA Detection 1:1000-1:10000
Monoclonal
Purified
RUO
Human
DSHPLYP, Duncan disease SH2-protein, EBVS, FLJ92177, IMD5, MTCP1, SAP/SH2D1A, SAPlymphoproliferative syndrome, SH2 domain containing 1A, SH2 domain-containing protein 1A, signaling lymphocyte activation molecule-associated protein, Signaling lymphocytic activation molecule-associated protein, SLAM associated protein/SH2 domain protein 1A, SLAM-associated protein, T cell signal transduction molecule SAP, T-cell signal transduction molecule SAP, XLPDFLJ18687, XLPSH2 domain protein 1A
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SH2D1A (RefSeq#: NP_002342.1; Pro2-Lys97).
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
13
ELISA, Sandwich ELISA Detection
Unconjugated
Mouse
Immune System Diseases, Immunology
0.2 μm filtered solution in PBS
4068
IgG
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.