missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SH2D1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30818-100ug
This item is not returnable.
View return policy
Description
SH2D1A Polyclonal antibody specifically detects SH2D1A in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| SH2D1A | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL, Immunoprecipitation | |
| DSHPLYP, Duncan disease SH2-protein, EBVS, FLJ92177, IMD5, MTCP1, SAP/SH2D1A, SAPlymphoproliferative syndrome, SH2 domain containing 1A, SH2 domain-containing protein 1A, signaling lymphocyte activation molecule-associated protein, Signaling lymphocytic activation molecule-associated protein, SLAM associated protein/SH2 domain protein 1A, SLAM-associated protein, T cell signal transduction molecule SAP, T-cell signal transduction molecule SAP, XLPDFLJ18687, XLPSH2 domain protein 1A | |
| Full-length recombinant SH2D1A protein | |
| 100 μg | |
| Immune System Diseases, Immunology | |
| 4068 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction