missing translation for 'onlineSavingsMsg'
Learn More

SH2D3A Rabbit anti-Human, Polyclonal, Proteintech

Product Code. 16814904 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16814904 20 μL 20µL
16804904 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16814904 Supplier Proteintech Supplier No. 156411AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

May play a role in JNK activation.

Specifications

Antigen SH2D3A
Applications Western Blot, Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SH2D3A
Gene Accession No. Q9BRG2
Gene Alias Novel SH2 containing protein 1, NSP1, SH2 domain containing 3A, SH2D3A
Gene Symbols SH2D3A
Host Species Rabbit
Immunogen SH2D3A Fusion Protein Ag8132
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10045
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.