missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3GL3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94877-0.1ml
This item is not returnable.
View return policy
Description
SH3GL3 Polyclonal antibody specifically detects SH3GL3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| SH3GL3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| CNSA3, EEN-B2, endophilin-A3, SH3D2C, SH3-domain GRB2-like 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human SH3GL3 (NP_003018.3). MSVAGLKKQFHKASQLFSEKISGAEGTKLDDEFLDMERKIDVTNKVVAEILSKTTEYLQPNPAYRAKLGMLNTVSKIRGQVKTTGYPQTEGLLGDCMLKYGKELGEDSTFGNALIEVGESMKLMAEVKDSLDINVKQTFIDPLQLLQDKDLKEIGHHLKKLEGRRLDYDYKKKRVGKIPDEEVRQAVEKFEESKELAERSMFNFLENDVEQVSQLAVFIEAALDYHRQSTEILQELQSKLQMRISAASSVPRREYKPRPVKRSSSELNGV | |
| 0.1 mL | |
| Neurodegeneration, Neuroscience, Signal Transduction | |
| 6457 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction